missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP22 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NLS6809
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DUSP22 Polyclonal antibody specifically detects DUSP22 in Human, Bovine, Equine, Primate, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
| DUSP22 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Bovine, Equine, Primate, Monkey | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 2 - 22 μg/mL | |
| dual specificity phosphatase 22, dual specificity protein phosphatase 22, EC 3.1.3.16, EC 3.1.3.48, FLJ35864, homolog of mouse dual specificity phosphatase LMW-DSP2, JKAP, JNK-stimulating phosphatase 1, JSP-1, JSP1JNK-stimulatory phosphatase-1, LMWDSP2, LMW-DSP2, Low molecular weight dual specificity phosphatase 2, Mitogen-activated protein kinase phosphatase x, MKP-x, MKPXMAP kinase phosphatase x, VHX | |
| Synthetic 17 aa peptide from near N-terminus of human DUSP22. | |
| 0.05 mL | |
| MAP Kinase Signaling, Protein Phosphatase | |
| 56940 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido