Learn More
VEE GEE Dual Scale Hydrometer
Descripción
- The Dual Scale Specific Gravity/Baume Hydrometer includes two scales.
- One scale is designed for liquids heavier than water.
- The other scale is specifically for liquids lighter than water.
Especificaciones
Especificaciones
| Tipo | Specific Gravity and Baume Hydrometer |
| Incrementos | 0.010/1° |
| Longitud (métrico) | 16.5 cm |
| Intervalo de Baume | De 0° a 41° (pesado) |
| Intervalo de gravedad específica | De 1,000 a 1,400 |
| Temperature of Standardization | 15.6°C (60°F) |