missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DTL Recombinant Rabbit Monoclonal Antibody (23GB5230), Invitrogen™

Descripción
Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control, DNA damage response and translesion DNA synthesis. The DCX(DTL) complex, also named CRL4(CDT2) complex, mediates the polyubiquitination and subsequent degradation of CDT1 and CDKN1A/p21(CIP1). CDT1 degradation in response to DNA damage is necessary to ensure proper cell cycle regulation of DNA replication. CDKN1A/p21(CIP1) degradation during S phase or following UV irradiation is essential to control replication licensing. Most substrates require their interaction with PCNA for their polyubiquitination: substrates interact with PCNA via their PIP-box, and those containing the 'K+4' motif in the PIP box, recruit the DCX(DTL) complex, leading to their degradation. In undamaged proliferating cells, the DCX(DTL) complex also promotes the 'Lys-164' monoubiquitination of PCNA, thereby being involved in PCNA-dependent translesion DNA synthesis.
Especificaciones
Especificaciones
| Antígeno | DTL |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 23GB5230 |
| Concentración | 1.6 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | DTL |
| N.º de referencia del gen | Q3TLR7, Q9NZJ0 |
| Alias de gen | 2810047L02Rik; 5730564G15Rik; CDT2; CDW1; DCAF2; DDB1 and CUL4 associated factor 2; DDB1- and CUL4-associated factor 2; denticleless E3 ubiquitin protein ligase homolog; denticleless E3 ubiquitin protein ligase homolog (Drosophila); denticleless homolog; denticleless homolog (Drosophila); denticleless protein homolog; DTL; L2dtl; lethal(2) denticleless protein homolog; Meth A RAMP; Meth A retinoic acid-regulated nuclear matrix-associated protein; RA regulated nuclear matrix associated protein; RAMP; RA-regulated nuclear matrix-associated protein; Retinoic acid-regulated nuclear matrix-associated protein; RGD1310439 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?