Learn More
BUCHI Lyovapor™ Drying Chamber
Código de producto. 15902711 Tienda Buchi Productos
Change view
Click to view available options
Tipo:
Benchtop Freeze Dryer
Heated Product Shelf Chamber
N.º de estantes:
0
4
6
Modelo:
12-position Manifold Top
24-position Manifold Top
Standard Top
Stoppering Vials Top
Incluye:
4 Sample Temperature Sensors
6 Sample Temperature Sensors
Chamber Only
Drying Chamber Only
Pressure Difference Testing
Tamaño de la unidad:
cada uno
Código de producto. Tipo N.º de estantes Modelo Incluye unitSize
15902711 Benchtop Freeze Dryer 6 Standard Top Drying Chamber Only cada uno
17254694 Heated Product Shelf Chamber 6 12-position Manifold Top 4 Sample Temperature Sensors cada uno
30130641 Heated Product Shelf Chamber 6 Standard Top Pressure Difference Testing cada uno
17234694 Heated Product Shelf Chamber 6 Standard Top 6 Sample Temperature Sensors cada uno
30130644 Heated Product Shelf Chamber 4 Stoppering Vials Top 4 Sample Temperature Sensors cada uno
17204694 Heated Product Shelf Chamber 4 Stoppering Vials Top Chamber Only cada uno
17284684 Heated Product Shelf Chamber 4 12-position Manifold Top 4 Sample Temperature Sensors cada uno
18804171 Heated Product Shelf Chamber 4 Standard Top Pressure Difference Testing cada uno
17264684 Heated Product Shelf Chamber 4 Standard Top Chamber Only cada uno
15912711 Benchtop Freeze Dryer 6 12-position Manifold Top Drying Chamber Only cada uno
30130646 Benchtop Freeze Dryer 0 Stoppering Vials Top Drying Chamber Only cada uno
15992701 Benchtop Freeze Dryer 4 12-position Manifold Top Drying Chamber Only cada uno
15982701 Benchtop Freeze Dryer 4 Standard Top Drying Chamber Only cada uno
15962701 Benchtop Freeze Dryer 0 24-position Manifold Top Drying Chamber Only cada uno
14 options
Código de producto. 15902711
Marca: BUCHI 11DCIN16100
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Freeze drying flexibility for basic and advanced drying of vials, shelves, and flasks.
  • Manifolds: adapters for traditional freeze drying beakers or round bottom flasks
  • Shelves: for vials, plates, and irregular shaped items
  • Heated Shelves: advanced control of drying process and applications
  • Stoppering Chambers: seal vials without exposure to ambient moisture
Specifica
Tipo Benchtop Freeze Dryer
N.º de estantes 6
Intervalo de temperatura (métrico) Ambient to -105°C
Material Acrílico
Modelo Standard Top
Incluye Drying Chamber Only
tipo de tapón Norteamérica
Tensión 220/240 V
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.