Learn More
DPY30 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 17264606 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17264606 20 μL 20 microlitros
17254606 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17264606 Proveedor Proteintech N.º de proveedor 162811AP20UL
Rabbit Polyclonal Antibody
The DPY30 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human DPY30. This antibody recognizes human, mouse, rat antigen. The DPY30 antibody has been validated for the following applications: IF, IHC,ELISA analysis.
Especificaciones
Antígeno DPY30
Aplicaciones Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Polyclonal
Concentración 400 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 84661
Especies diana Human, Mouse, Rat
Formulario Liquid
Isotype IgG