missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPP10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13434399
Change view
Click to view available options
Cantidad:
50 μg
Unit Size:
50 microgramos
Product Code. Cantidad unitSize
13434399 50 μg 50 microgramos
1 options
This item is not returnable. View return policy

Product Code. 13434399

Brand: Invitrogen™ PA532725

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Bovine, Panda, Horse, Rabbit, Pig (100%); Gibbon, Dog (93%); Bat, Elephant (87%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

A novel gene, DPP10, encodes a homolog of dipeptidyl peptidases (DPPs) that cleave terminal dipeptides from cytokines and chemokines, and it presents a potential new target for asthma therapy. The DPP10 protein shares features with members of the S9B family of DPP serine proteases, which includes DPP4, a widely expressed enzyme that plays a central role in chemokine processing as part of the innate immune system. The locus displays a complex pattern of transcript splicing, with eight alternate first axons; four of which localize within the LD block strongly associated with asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno DPP10
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.1% sodium azide
génica DPP10
N.º de referencia del gen Q8N608
Alias de gen 6430601K09Rik; dipeptidyl peptidase 10; dipeptidyl peptidase IV-related protein 3; dipeptidyl peptidase like 10; dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; dipeptidylpeptidase 10; dipeptidyl-peptidase 10 (inactive); dipeptidyl-peptidase 10 (non-functional); DPL2; DPP X; DPP10; DPPY; Dprp3; DPRP-3; HGNC:20823; inactive dipeptidyl peptidase 10; KIAA1492; Kv4 potassium channel auxiliary subunit; OTTHUMP00000203618
Símbolos de los genes DPP10
Especie del huésped Rabbit
Inmunógeno Synthetic 15 amino acid peptide from extracellular domain of human DPP10.
Método de purificación Antigen Affinity Chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100053255, 100157195, 100346104, 57628, 617222
Especies diana Human, Rabbit, Monkey, Bovine, Pig, Horse
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.