Learn More
Invitrogen™ DP1 Recombinant Rabbit Monoclonal Antibody (24GB2685)
GREENER_CHOICE
Código de producto. 30275530
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30275530 100 μL 100 microlitros
30276455 20 μL 20 microlitros
2 options
Código de producto. 30275530
Marca: Invitrogen™ MA557794
Rabbit Recombinant Monoclonal Antibody
DP1 Recombinant Monoclonal Antibody for Western Blot, Flow
DP family contains at least two members, DP1 and 2. Structurally, DP1 and 2 are relatively conserved, yet their expression appears to be tissue specific. E2F 1, a functional target for the tumor suppressor protein Rb, heterodimerizes with DP1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno DP1
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB2685
Concentración 0.353 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica TFDP1
N.º de referencia del gen Q08639, Q14186
Alias de gen AS1; ASRT1; cell cycle regulatory transcription factor DP1; DILC; down-regulated in liver cancer stem cells; DP; DP1; Dp-1; DRTF1; DRTF1-polypeptide 1; E2F dimerization partner 1; E2F-related transcription factor; MGC49004; OTTHUMP00000018767; Tfdp1; transcription factor Dp 1; transcription factor Dp-1
Símbolos de los genes LOC100360176, TFDP1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human DP1 (aa 351-410).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100360176, 21781, 361178, 7027
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG