Learn More
Melford Doxycycline HCl
Descripción
Doxycycline HCl is a widely used antibiotic in the life science marketplace.
It belongs to the tetracycline class of antibiotics and exhibits broad-spectrum activity against both gram-positive and gram-negative bacteria, as well as certain other microorganisms.
In the life science marketplace, Doxycycline HCl finds applications in various fields, including research and biotechnology. It is commonly used as an antibacterial agent in cell culture systems, where it helps prevent bacterial contamination in cell lines and maintain sterile conditions.
Doxycycline HCl is also utilized in molecular biology techniques such as gene expression studies and protein production. It can be added to media to select for cells containing specific genes or recombinant DNA constructs, as it inhibits protein synthesis in bacteria and other organisms. Its ability to induce gene expression or regulate protein production is exploited in certain research applications.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Doxycycline HCl |
| CAS | 10592-13-9 |
| Cantidad | 100 g |
| Fórmula molecular | C22H24N2O8-HCl |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.