Learn More
Invitrogen™ Donkey anti-Mouse IgG (H+L) ReadyProbes™ Secondary Antibody, Alexa Fluor™ 488
Código de producto. 15136896
Change view
Click to view available options
Cantidad:
2 x 2.5 mL
Tamaño de la unidad:
2 ampollas
15136896 2 x 2.5 mL 2 ampollas
1 options
Código de producto. 15136896
Marca: Invitrogen™ R37114
Donkey Polyclonal Secondary Antibody
No pipetting or dilutions required. Just tip and drip two drops per mL buffer to stain your cells. This antibody shows minimum cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat, and sheep serum proteins. Like other ReadyProbes reagents, Alexa Fluor 488 Donkey anti-Mouse IgG Antibody comes in a convenient dropper bottle.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Mouse IgG (H+L)
Aplicaciones Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Conjugado Alexa Fluor 488
Formulación PBS with 0.1% BSA and <0.1% sodium azide
Especie del huésped Donkey
Inmunógeno Gamma Immunoglobin.
Método de purificación Affinity chromatography
Cantidad 2 x 2.5 mL
Estado normativo RUO
Primario o secundario Secondary
Especies diana Mouse
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG
Características ecológicas Responsibly packaged