missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Donkey anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 594
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, mouse, rabbit, rat and sheep.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antígeno | Human IgG (H+L) Cross-Adsorbed |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | DyLight 594 |
| Formulación | PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 |
| Especie del huésped | Donkey |
| Inmunógeno | Human IgG-heavy and light chain. |
| Método de purificación | Antigen affinity chromatography |
| Cantidad | 500 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?