missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Donkey anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 550
Descripción
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, mouse, rabbit, rat and sheep.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Especificaciones
Especificaciones
| Antígeno | Human IgG (H+L) Cross-Adsorbed |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | DyLight 550 |
| Formulación | PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 |
| Especie del huésped | Donkey |
| Inmunógeno | Human IgG-heavy and light chain. |
| Método de purificación | Antigen affinity chromatography |
| Cantidad | 500 μg |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?