Learn More
Invitrogen™ Donkey anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 488
Código de producto. 13477277
Change view
Click to view available options
Cantidad:
500 μg
Tamaño de la unidad:
500 microgramos
13477277 500 μg 500 microgramos
1 options
Código de producto. 13477277 Proveedor Invitrogen™ N.º de proveedor SA510126
Donkey Polyclonal Secondary Antibody
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, mouse, rabbit, rat and sheep.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Human IgG (H+L) Cross-Adsorbed, Human IgG (H+L)
Aplicaciones Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado DyLight 488
Formulación PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Especie del huésped Donkey
Inmunógeno Human IgG-heavy and light chain.
Método de purificación Antigen affinity chromatography
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Human
Contenido y almacenamiento 4°C
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG