Learn More
DOK7 Mouse anti-Human, Clone: OTI3C12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF504832
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is essential for neuromuscular synaptogenesis. The protein functions in aneural activation of muscle-specific receptor kinase, which is required for postsynaptic differentiation, and in the subsequent clustering of the acetylcholine receptor in myotubes. This protein can also induce autophosphorylation of muscle-specific receptor kinase. Mutations in this gene are a cause of familial limb-girdle myasthenia autosomal recessive, which is also known as congenital myasthenic syndrome type 1B. Alternative splicing results in multiple transcript variants.Specifications
| DOK7 | |
| Monoclonal | |
| Unconjugated | |
| DOK7 | |
| C4orf25, CMS10, CMS1B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 285489 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI3C12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q18PE1 | |
| DOK7 | |
| Full length human recombit protein of human DOK7 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.