missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14087-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.Especificaciones
| DOK2 | |
| Polyclonal | |
| Unconjugated | |
| DOK2 | |
| Docking protein 2, docking protein 2, 56kDa, DOK2, p56(dok 2), p56DOK, p56dok 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13449, 290361, 9046 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60496, O70469 | |
| Dok2 | |
| DOK2 Fusion Protein Ag5218 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido