missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-93647-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DOCK3 Polyclonal specifically detects DOCK3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| DOCK3 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q8IZD9 | |
| DOCK3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FEVVDSDQISVSDLYKMHLSSRQSVQQSTSQVDTMRPRHGETCRMPVPHHFFLSLKSFTYNTIGEDTDVFFSLYDMREGKQISERFLVRLNKNG | |
| Affinity Purified | |
| RUO | |
| 1795 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| dedicator of cytokinesis 3, KIAA0299, modifier of cell adhesion | |
| Rabbit | |
| 233 kDa | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido