missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNase I Polyclonal Antibody

Descripción
DNase I Polyclonal Antibody for Western Blot, IHC (P)
Internucleosomal DNA fragmentation following the activation of endonucleases is the common end point of apoptosis. DNase I, a Ca2+/Mg2+-dependent endonuclease ubiquitously expressed in mammalian tissues, has been implicated to mediate internucleosomal DNA degradation in human cells undergoing apoptosis. DNase I is highly polymorphic, and at least six alleles of DNase I are known. DNase II, the ubiquitously expressed acidic deoxyribonuclease, acts downstream of caspase activation and may also induce DNA digestion during apoptosis. DNase I cleaves DNA to 5'-phosphodinucleotide and 5'-phospho- oligonucleotide end-products, whereas DNase II cleaves DNA to 3'-phosphomononucleotide and 3'-phosphooligonucleotide end-products. The mechanism by which DNase II cuts DNA is similar to DNase I, which produces nicks rather than double-strand cuts. DNase II is usually present in cytoplasm of epithelial cells, but it appears concentrated in the nuclei of lens fibers. In contrast, DNase I is always concentrated in nuclei of epithelial and fiber cells.
Especificaciones
Especificaciones
| Antígeno | DNase I |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | DNASE1 |
| N.º de referencia del gen | P24855, P49183 |
| Alias de gen | AI788650; deoxyribonuclease; Deoxyribonuclease 1; deoxyribonuclease I; Deoxyribonuclease1; Deoxyribonuclease-1; DeoxyribonucleaseI; DNASE 1; DNase I; DNase I lysosomal; DNase I, lysosomal; Dnase1; DNaseI; DNL 1; DNL1; Dornase alfa; DRNI; epithelial lens deoxyribonuclease I; FLJ38093; human urine deoxyribonuclease I; pancreatic deoxyribonuclease I |
| Símbolos de los genes | DNASE1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?