missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25444-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DjC9 (DnaJ subfamily C member 9) belongs to the Hsp40 (heat shock protein 40) family of proteins that function as co-chaperones to Hsp70.Hsp40 proteins are classified into 3 main subfamilies (A, B, and C). The families are defined by the presence of particular domains which include a highly conserved alpha-helical N-terminal domain termed the J-domain, a glycine/phenylalanine-rich region, a cysteine-rich region, and a C-terminal beta-sheet containing domain. DjC9 is part of subfamily C that bears only the J-domain. DjC9 has been shown to interact with HSP70 through its J domain and activate its ATPase activity.Especificaciones
| DNAJC9 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC9 | |
| DnaJ protein SB73, DNAJC9, HDJC9, JDD1, KIAA0974, SB73 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23234 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WXX5 | |
| DNAJC9 | |
| DNAJC9 Fusion Protein Ag22123 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido