missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJB9 Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA5142100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000. Preliminary analysis showed no signal in Western Blot.
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
Especificaciones
| DNAJB9 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB9 | |
| AA408011; AA673251; AA673481; AW556981; DKFZP564F1862; DNA j protein b9; DnaJ (Hsp40) homolog, subfamily B, member 9; DnaJ heat shock protein family (Hsp40) member B9; dnaJ homolog subfamily B member 9; dnaJ homolog, subfamily b, member 9; DNAJB9; Endoplasmic reticulum DNA J domain-containing protein 4; endoplasmic reticulum DnaJ homolog 4; ERdj4; ER-resident protein ERdj4; Mdg1; MDG-1; mDj7; microvascular endothelial differentiation gene 1; microvascular endothelial differentiation gene 1 protein; MST049; MSTP049; UNQ743/PRO1471 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4189 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UBS3 | |
| DNAJB9 | |
| Peptide with sequence RGNMVTTYTDCSGQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido