missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJB4 Polyclonal Antibody
Descripción
Immunogen sequence: EAYEVLSDPK KREIYDQFGE EGLKGGAGGT DGQGGTFRYT FHGDPHATFA AFFGGSNPFE IFFGRRMGGG RDSEEMEIDG DPFSAFGFSM NGYPRDRNSV GPSRLKQDPP V.
DnaJ heat shock induced proteins are from the bacterium Escherichia coli and are under the control of the htpR regulatory protein. The DnaJ proteins play a critical role in the HSP 70 chaperone machine by interacting with HSP 70 to stimulate ATP hydrolysis. The proteins contain cysteine rich regions that are composed of zinc fingers that form a peptide binding domain responsible for the chaperone function. DnaJ proteins are important mediators of proteolysis and are involved in the regulation of protein degradation, exocytosis and endocytosis. DnaJB4 (DnaJ homolog subfamily B member 4), also known as HLJ1, is expressed in skeletal muscle, heart and pancreas, and lower expression in brain, placenta and liver.
Especificaciones
Especificaciones
| Antígeno | DNAJB4 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.34 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| génica | Dnajb4 |
| N.º de referencia del gen | Q9D832, Q9UDY4 |
| Alias de gen | 1700029A20Rik; 2010306G19Rik; 5730460G06Rik; DjB4; DnaJ (Hsp40) homolog, subfamily B, member 4; DnaJ (Hsp40) homolog, subfamily B, member 4-like; DnaJ heat shock protein family (Hsp40) member B4; dnaJ homolog subfamily B member 4; DNAJB 4; Dnajb4; DnaJ-like heat shock protein 40; DNAJW; Heat shock 40 kDa protein 1 homolog; heat shock protein 40 homolog; HLJ 1; HLJ1; HSP40 homolog; human liver DnaJ-like protein |
| Símbolos de los genes | Dnajb4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?