missing translation for 'onlineSavingsMsg'
Learn More

DMRT1 Monoclonal Antibody (OTI4E12), TrueMAB™, OriGene

Código de producto. p-200015897
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
17598343 30 μL 30 microlitros
17578343 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17598343 Supplier Origene Technologies Supplier No. TA807669S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DMRT1 Monoclonal antibody specifically detects DMRT1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antígeno DMRT1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon OTI4E12
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica Kctd9
N.º de referencia del gen Q7L273
Alias de gen CT154; DM domain expressed in testis 1; DM domain expressed in testis protein 1; Dmrt1; DMT1; Doublesex and mab-3 related transcription factor 1; doublesex- and mab-3-related transcription factor 1; Unknown (protein for MGC:134119)
Especie del huésped Mouse
Inmunógeno Human recombinant protein fragment corresponding to amino acids 1-223 of human KCTD9 produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 54793
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.