missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-3-Indolylglycine, MedChemExpress
Description
DL-3-Indolylglycine is an unnatural amino acid that is very similar to Tryptophan, with the indole moiety directly attached to the α-position.
Specifications
Specifications
| Nombre del producto químico o material | DL-3-Indolylglycine |
| CAS | 6747-15-5 |
| Color | Off-White |
| Notas de grado de pureza | Research |
| Fórmula molecular | C10H10N2O2 |
| Información de solubilidad | H2O : 4.63 mg/mL (24.34 mM; ultrasonic and warming and adjust pH to 2 with HCl and heat to 60°C) |
| SMILES | NC(C(O)=O)C1=CNC2=CC=CC=C21 |
| Peso molecular (g/mol) | 190.2 |
| Cantidad | 500 mg |
| Formula Weight (peso de la fórmula) | 190.2 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?