Learn More
BOLA Distillation Apparata
Código de producto. 10629601
Change view
Click to view available options
Capacidad (métrico):
500 mL
1000 mL
2000 mL
4000 mL
6000 mL
Tamaño de la unidad:
cada uno
Código de producto. Capacidad (métrico) unitSize
10629601 6000 mL cada uno
10319231 500 mL cada uno
10104491 1000 mL cada uno
10247451 2000 mL cada uno
10787302 4000 mL cada uno
5 options
Código de producto. 10629601
Marca: BOLA B28015
Suitable for the distillation of strong alkaline or acid products as well as very aggressive solvents.
  • Distillation Apparata of PTFE - with SAFE LAB-System consisting of: Reaction vessel
  • flat flange (B281-03) Gasket
  • flat flange (B 282-02) Lid
  • flat flange (B 283-02) Joining piece
  • flat flange (B 284-02) Dropping funnel
  • 125 ml (B 285-01) Supporting device for thermometer (B 286-03)
  • Thermometer for flasks (B 287-03)
  • Stirrer bearing (B288-02)
  • Moon-Shaped stirrer shaft (C 376-12)
  • Stillhead 2x NS 29/32 (B 289-03)
  • Distillation thermometer (B 290-03)
  • Liebig condenser 270 mm (B 291-02)
  • Receiver adapter (B 292-02)
  • Vacuum stopcock (B 293-02)
  • Round-bottom flask (A 158-06)
Especificaciones
Capacidad (métrico) 6000 mL
Dimensiones (Largo x Ancho x Alto) 790 x 1000 mm
Esterilizables en autoclave Autoclavable