Learn More
BOLA Distillation Apparata
Código de producto. 10247451
Click to view available options
Capacidad (métrico):
500 mL
1000 mL
2000 mL
4000 mL
6000 mL
Tamaño de la unidad:
cada uno
Código de producto. 10247451
Marca: BOLA B28009
Suitable for the distillation of strong alkaline or acid products as well as very aggressive solvents.
  • Distillation Apparata of PTFE - with SAFE LAB-System consisting of: Reaction vessel
  • flat flange (B281-03) Gasket
  • flat flange (B 282-02) Lid
  • flat flange (B 283-02) Joining piece
  • flat flange (B 284-02) Dropping funnel
  • 125 ml (B 285-01) Supporting device for thermometer (B 286-03)
  • Thermometer for flasks (B 287-03)
  • Stirrer bearing (B288-02)
  • Moon-Shaped stirrer shaft (C 376-12)
  • Stillhead 2x NS 29/32 (B 289-03)
  • Distillation thermometer (B 290-03)
  • Liebig condenser 270 mm (B 291-02)
  • Receiver adapter (B 292-02)
  • Vacuum stopcock (B 293-02)
  • Round-bottom flask (A 158-06)
Especificaciones
Capacidad (métrico) 2000 mL
Dimensiones (Largo x Ancho x Alto) 700 x 750 mm
Esterilizables en autoclave Autoclavable