Learn More
Poulten Graf™ Adaptador para dispensador de PTFE blanco GL38 Optifix 101.091-38
Descripción
For strong acids (excl. HF) and aggressive chemicals. Visible dispensing process, dispensing piston made of PTFE-sleeved, solid glass, with additional sealing lip at the end of the piston to prevent acid fumes and odours from escaping.