Learn More
Dipenteno, téc., Thermo Scientific Chemicals
Código de producto. p-7059959
Change view
Click to view available options
Cantidad:
250 mL
Tamaño de la unidad:
250 ml
15422457 250 mL 250 ml
1 options
Código de producto. 15422457
Marca: Thermo Scientific Alfa Aesar 045805.AK
Solicitar formato a granel o personalizado
Dipentene is a monoterpene olefin having potential applications in polymer and fuel chemistry. It is also widely used as flavoring and fragrance agent. It is a promising green solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 138-86-3
Fórmula molecular C10H16
Peso molecular (g/mol) 136.24
Número MDL MFCD00062992
Clave InChI XMGQYMWWDOXHJM-UHFFFAOYNA-N
Sinónimo dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen
PubChem CID 22311
ChEBI CHEBI:15384
SMILES CC(=C)C1CCC(C)=CC1
Especificaciones
Nombre del producto químico o material Dipentene
Densidad 0.85 g/mL
Punto de ebullición 170°C to 180°C
Temperatura de inflamación 43°C (109°F)
Fórmula lineal CH3C6H8C(CH3)=CH2
Índice de refracción 1.474
Cantidad 250 mL
Número UN UN2052
Beilstein 3587825
Índice Merck 14,5493
Información de solubilidad Soluble in ether (miscible) and ethanol (miscible). Insoluble in water.
Nombre IUPAC 1-metil-4-prop-1-en-2-ilciclohexeno
Formula Weight (peso de la fórmula) 136.24
Grado Técnico
Forma física Líquida