missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Digoxigenin Polyclonal Antibody

Descripción
Sheep anti digoxigenin antibody recognizes digoxigenin, a steroid found in plants that is often attached to oligonucleotides and used as a molecular probe to detect mRNA production.
Digoxin, also known as digitalis, is a purified cardiac glycoside extracted from the foxglove plant, Digitalis lanata. Its corresponding aglycone is digoxigenin, and its acetyl derivative is acetyldigoxin. Digoxin is widely used in the treatment of various heart conditions, namely atrial fibrillation, atrial flutter and sometimes heart failure that cannot be controlled by other medication. Digoxin preparations are commonly marketed under the trade names Lanoxin, Digitek, and Lanoxicaps. Digoxin is also used as an affinity system for nonradioactive labeling and detection of proteins and nucleic acids.
Especificaciones
Especificaciones
| Antígeno | Digoxigenin |
| Aplicaciones | ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.2 |
| Alias de gen | C23H34O5; DIG; Digitalis |
| Especie del huésped | Sheep |
| Inmunógeno | Digoxin-3-CM conjugated to KLH. |
| Método de purificación | Protein G |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?