missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB7520R-01M
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Digoxigenin Monoclonal specifically detects Digoxigenin in Multi-Species samples. It is validated for Immunohistochemistry.
Especificaciones
| Digoxigenin | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects Digoxigenin labeled proteins, nucleic acids and Digoxigenin conjugated primary and secondary antibodies. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 611621R | |
| Immunohistochemistry 5-25 ug/mL | |
| DIG | |
| KLH-coupled Digoxigenin | |
| 1 mg | |
| Primary | |
| Multi-species | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido