Learn More
Faithful™ Differential Blood Cell Counter
Descripción
- ABS outer shell can resist collision.
- Color charts of white blood cells are supplied.
- Bell sound at 100% total record count.
- Reset knob clears figures.
- Quality certification: ISO 9001, ISO 13485
Especificaciones
Especificaciones
| N.º de llaves | 5 Keys |
| Dimensiones (Largo x Ancho x Alto) | 250 x 80 x 50 mm |
| Incluye | Totalizer |