Learn More
Polvo de diamante, natural, 40-60 micrómetros, 99,9 % (base metálica), Thermo Scientific Chemicals
Descripción
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Diamond powder |
| Nombre de nota | Natural |
| Forma física | Polvo |
| Cantidad | 5 g |
| Notas del porcentaje del ensayo | (base metálica) |
| Número MDL | MFCD00211867 |
| Porcentaje de pureza | 99.9% |