missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: ISHAKDKLVY VHTNGPKKKK VTLHIKWPKS VEVEGYGSKK IDAERQAAAA ACQLFKGWGL LGPRNELFDA AKYRVLADRF GSPA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DHX30 is a member of this family.DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The encoded protein has 97% sequence identity with the mouse HELG protein. Alternative splicing of this gene generates 3 transcript variants.
Specifications
Specifications
| Antígeno | DHX30 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | DHX30 |
| N.º de referencia del gen | Q7L2E3 |
| Alias de gen | 2810477H02Rik; ATP-dependent RNA helicase DHX30; ATP-dependent RNA helicase DHX30; putative ATP-dependent RNA helicase DHX30; C130058C04Rik; Ddx30; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30; DEAH (Asp-Glu-Ala-His) box helicase 30; DEAH (Asp-Glu-Ala-His) box polypeptide 30; DEAH box protein 30; DEAH-box helicase 30; DExH-box helicase 30; Dhx30; FLJ11214; Helg; KIAA0890; putative ATP-dependent RNA helicase DHX30; RETCOR; Ret-CoR; retina co-repressor; Unknown (protein for MGC:134480) |
| Símbolos de los genes | DHX30 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
