Learn More
DHX16 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16887803 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16887803 20 μL 20 microlitros
16877803 150 μL 150 microlitros
2 options
Código de producto. 16887803
Marca: Proteintech 110211AP20UL
Rabbit Polyclonal Antibody
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a functional homolog of fission yeast Prp8 protein involved in cell cycle progression. This gene is mapped to the MHC region on chromosome 6p21.3, a region where many maligt, genetic and autoimmune disease genes are linked. Three transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Antígeno DHX16
Aplicaciones Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica DHX16
N.º de referencia del gen O60231
Alias de gen ATP dependent RNA helicase 3, DBP2, DDX16, DEAH box protein 16, DHX16, KIAA0577, PRO2014, PRP8
Símbolos de los genes DHX16
Especie del huésped Rabbit
Inmunógeno DHX16 Fusion Protein Ag1482
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 69192, 8449
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG