missing translation for 'onlineSavingsMsg'
Learn More

(+)-DHMEQ, MedChemExpress

Código de producto. 16488717
Encompass_Preferred
Change view
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10 miligramos
2 miligramos
25 miligramos
5 miligramos
Product Code. Cantidad unitSize
16488717 5 mg 5 miligramos
16496118 10 mg 10 miligramos
16490216 2 mg 2 miligramos
16443436 25 mg 25 miligramos
4 options
This item is not returnable. View return policy

Product Code. 16488717

Brand: Medchem Express HY14645A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress (+)-DHMEQ is an activator of antioxidant transcription factor Nrf2. (+)-DHMEQ is the enantiomer of (-)-DHMEQ. (-)-DHMEQ inhibits NF-kB than its enantiomer (+)-DHMEQ.

(+)-DHMEQ is an activator of antioxidant transcription factor Nrf2. (+)-DHMEQ is the enantiomer of (-)-DHMEQ. (-)-DHMEQ inhibits NF-kB than its enantiomer (+)-DHMEQ.

Specifications

Nombre del producto químico o material (+)-DHMEQ
CAS 287194-41-6
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C13H11NO5
Cantidad 5 mg
Sinónimo (1R,2R,6R)-Dehydroxymethylepoxyquinomicin (1R,2R,6R)-DHMEQ
Información de solubilidad DMSO : 100 mg/mL (382.80 mM; Need ultrasonic)
SMILES O=C(NC([C@@H](O)[C@@]1([H])O[C@@]21[H])=CC2=O)C3=CC=CC=C3O
Peso molecular (g/mol) 261.23
Formula Weight (peso de la fórmula) 261.23
Porcentaje de pureza 99.65%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.