Learn More
Invitrogen™ DHFR Recombinant Rabbit Monoclonal Antibody (23GB3575), Invitrogen™
GREENER_CHOICE
Código de producto. 30089049
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30089049 100 μL 100 microlitros
30087413 20 μL 20 microlitros
2 options
Código de producto. 30089049
Marca: Invitrogen™ MA553398
Rabbit Recombinant Monoclonal Antibody
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno DHFR
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 23GB3575
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica DHFR
N.º de referencia del gen P00374, P00375, Q920D2
Alias de gen 8430436I03Rik; AA607882; AI662710; AW555094; Dhfr; Dhfr1; DHFRP1; Dihydrofolate reductase; Dihydrofolate reductase 1 (active); DYR
Símbolos de los genes DHFR
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human DHFR (134-187AA).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13361, 1719, 24312
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG