missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHDH Polyclonal Antibody

Descripción
Recommended positive controls: DHDH tansfected 293T cell. Predicted reactivity: Dog (84%), Pig (82%), Rabbit (82%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction.
Especificaciones
Especificaciones
| Antígeno | DHDH |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.99 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| génica | DHDH |
| N.º de referencia del gen | Q9UQ10 |
| Alias de gen | 1300018L09Rik; 2DD; 3-deoxyglucosone reductase; AA591799; CAN2DD; ch211-203b17.3; DDH; DHDH; dhdh.2; dhdh-like.1.L; dihydrodiol dehydrogenase; dihydrodiol dehydrogenase (dimeric); dihydrodiol dehydrogenase, tandem duplicate 2; dimeric dihydrodiol dehydrogenase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; Hum2DD; putative LOC617564 protein; SUS2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase L homeolog; XELAEV_18036112mg; Xld1; Xld-1; xylose dehydrogenase 1; zgc:101723 |
| Símbolos de los genes | DHDH |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?