missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGKG Monoclonal Antibody (613531)

Descripción
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes an enzyme that is a member of the type I subfamily of diacylglycerol kinases, which are involved in lipid metabolism. These enzymes generate phosphatidic acid by catalyzing the phosphorylation of diacylglycerol, a fundamental lipid second messenger that activates numerous proteins, including protein kinase C isoforms, Ras guanyl nucleotide-releasing proteins and some transient receptor potential channels. Diacylglycerol kinase gamma has been implicated in cell cycle regulation and in the negative regulation of macrophage differentiation in leukemia cells. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | DGKG |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 613531 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with trehalose and no preservative |
| génica | DGKG |
| N.º de referencia del gen | P49619, P49620, Q91WG7 |
| Alias de gen | 2900055E17Rik; 88 kDa diacylglycerol kinase; 90kDa; AI854428; DAG kinase gamma; Dagk3; DGKG; DGK-gamma; Diacylglycerol kinase gamma; diacylglycerol kinase, gamma; diacylglycerol kinase, gamma 3; diacylglycerol kinase, gamma 90kDa; diacylglyerol kinase gamma; diglyceride kinase gamma; E430001K23Rik; mKIAA4131 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?