missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DFNB31 Polyclonal Antibody

Descripción
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene is thought to function in the organization and stabilization of sterocilia elongation and actin cystoskeletal assembly, based on studies of the related mouse gene. Mutations in this gene have been associated with autosomal recessive non-syndromic deafness and Usher Syndrome. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
Especificaciones
Especificaciones
| Antígeno | DFNB31 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | WHRN |
| N.º de referencia del gen | Q80VW5, Q9P202 |
| Alias de gen | 1110035G07Rik; autosomal recessive deafness type 31 protein; AW122018; AW742671; C430046P22Rik; CASK-interacting protein CIP98; CIP98; deafness, autosomal recessive 31; Dfnb31; KIAA1526; PDZD7B; USH2D; whirler; whirlin; Whrn; wi |
| Símbolos de los genes | WHRN |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?