Learn More
Dextrina, precipitada por alcohol, Thermo Scientific Chemicals
Código de producto. p-7022802
Change view
Click to view available options
Cantidad:
100 g
500 g
Tamaño de la unidad:
100 g
500 g
11477697 100 g 100 g
11487697 500 g 500 g
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11477697
missing translation for 'mfr': Thermo Scientific Alfa Aesar A15717.22
Solicitar formato a granel o personalizado
This item is not returnable. View return policy
CAS: 9004-53-9 | C18H32O16 | 504.438 g/mol
Dextrin is used as adhesive in the manufacture of gummed tapes, textiles and paper. It is used in producing nutritional products. It is also used as a absorbents, binding agents and viscosity controlling agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9004-53-9
Fórmula molecular C18H32O16
Peso molecular (g/mol) 504.438
Número MDL MFCD00081554
Clave InChI FYGDTMLNYKFZSV-MRCIVHHJSA-N
Sinónimo dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
PubChem CID 62698
Nombre IUPAC (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihidroxi-2-(hidroximetil)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihidroxi-2-(hidroximetil)oxano-3-il]oxioxan-3-il]oxi-6-(hidroximetil)oxano-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
Specifications
Nombre del producto químico o material Dextrin
Nombre de nota precipitated by alcohol
Índice Merck 14,2953
Información de solubilidad Soluble in hot water (0.1g/ml).
Cantidad 100 g
Sensibilidad Hygroscopic