Learn More
DDX58 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16830125 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16830125 20 μL 20 microlitros
16820125 150 μL 150 microlitros
2 options
Código de producto. 16830125
Marca: Proteintech 205661AP20UL
Rabbit Polyclonal Antibody
The antibody is specific to DDX58.
Retinoic acid-inducible gene I, RIG-I is a pattern recognition receptor (PRR) involved in the recognition of viral dsRNA. Along with MDA5, RIG-I detects viral dsRNA and activates the innate immune response. Both MDA5 and RIG-I are RNA helicases and they perform overlapping as well as distinct roles. RIG-I is activated by dsRNAs without a 5′-triphosphate end and short dsRNAs, whereas MDA5 is activated by long dsRNAs. Once activated, both proteins signal through IPS-1 activating transcription factors NF-kappaB and IRF-3 (1) and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus (3), paromyxoviruses, Japanese encephalitis virus, and West Nile virus. MicroRNA-146a has been implicated in feedback inhibition of RIG-I-dependant antiviral response by negatively regulating RIG-I targets TRAF6, IRAK1, and IRAK2. Recent evidence has implicated RIG-I in the detection of cytosolic DNA through RNA polymerase III activity.
Especificaciones
Antígeno DDX58
Aplicaciones Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.51 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica DDX58
N.º de referencia del gen O95786, Q6Q899
Alias de gen DDX58, DEAD box protein 58, RIG 1, RIG I, RIG-I
Símbolos de los genes Ddx58
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 230073, 23586
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG