missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX3 Monoclonal Antibody (6G8)

Código de producto. 16396374 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
16396374 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16396374

Marca: Invitrogen™ MA532971

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene is a member of the large DEAD-box protein family, that is defined by the presence of the conserved Asp-Glu-Ala-Asp (DEAD) motif, and has ATP-dependent RNA helicase activity. This protein has been reported to display a high level of RNA-independent ATPase activity, and unlike most DEAD-box helicases, the ATPase activity is thought to be stimulated by both RNA and DNA. This protein has multiple conserved domains and is thought to play roles in both the nucleus and cytoplasm. Nuclear roles include transcriptional regulation, mRNP assembly, pre-mRNA splicing, and mRNA export. In the cytoplasm, this protein is thought to be involved in translation, cellular signaling, and viral replication. Misregulation of this gene has been implicated in tumorigenesis. This gene has a paralog located in the nonrecombining region of the Y chromosome. Pseudogenes sharing similarity to both this gene and the DDX3Y paralog are found on chromosome 4 and the X chromosome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno DDX3
Aplicaciones Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot
Clasificación Monoclonal
Clon 6G8
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
génica DDX3X
N.º de referencia del gen O00571, Q62167
Alias de gen ATP-dependent RNA helicase DDX3X; CAP-Rf; D1Pas1-related sequence 2; D1Pas1-rs2; DBX; DDX14; DDX3; Ddx3x; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 3; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 3; DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked; DEAD box protein 3, X-chromosomal; DEAD box RNA helicase DEAD3; DEAD box, X isoform; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, X-linked; DEAD/H box-3; Dead3; DEAD-box helicase 3, X-linked; DEAD-box protein 3 (DEAD-box RNA helicase DEAD3) (mDEAD3) (Embryonic RNA helicase) (D1PAS1 related sequence 2); Embryonic RNA helicase; Erh; fibroblast growth factor inducible 14; Fin14; Helicase-like protein 2; HLP2; mDEAD3; MRX102
Símbolos de los genes DDX3X
Especie del huésped Mouse
Inmunógeno Purified recombinant human DDX3 protein fragments expressed in E.coli.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13205, 1654, 317335
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.