missing translation for 'onlineSavingsMsg'
Learn More

DCT Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Código de producto. p-7205022 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
16841144 20 μL 20 microlitros
16831144 150 μL 150 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16841144

Marca: Proteintech 130951AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The TYR-related proteins, TRP1 and TRP2, are also specifically expressed in melanocytes, and they likewise contribute to the synthesis of melanin within the melanosomes. The TRPs, including TYR, all share a similar transmembrane region, contain two metal-binding regions and a cysteinerich epidermal growth factor motif, and are localized in the melanosomal membrane. These proteins, however, have distinct catalytic activity, and they individually contribute to the biosynthesis of melanin biopolymers. The TRPs are believed to exist as a multi-enzyme complex, as these proteins form aggregates together, and the expression of TRP1 also helps stabilize TYR in melanocytes.

Specifications

Antígeno DCT
Aplicaciones Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica DCT
N.º de referencia del gen P29812, P40126
Alias de gen DCT, DT, L dopachrome Delta isomerase, L dopachrome tautomerase, TRP 2, TRP2, Tyrosinase related protein 2, TYRP2
Símbolos de los genes Dct
Especie del huésped Rabbit
Inmunógeno DCT Fusion Protein Ag3714
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13190, 1638, 290484
Especies diana Mouse, Non-human Primate, Rat, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.