missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DcR3 Polyclonal Antibody

Descripción
DcR3 Polyclonal Antibody for Western Blot, IHC (P), Flow
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
Especificaciones
Especificaciones
| Antígeno | DcR3 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | TNFRSF6B |
| N.º de referencia del gen | O95407 |
| Alias de gen | DcR3; Decoy receptor 3; decoy receptor 3 variant 1; decoy receptor 3 variant 2; decoy receptor for Fas ligand; DJ583P15.1.1; hCG_2044836; M68; M68E; OTTHUMP00000031583; TNF receptor superfamily member 6b; TNFRSF6B; TR6; tumor necrosis factor receptor superfamily member 6B; tumor necrosis factor receptor superfamily member 6b, decoy; tumor necrosis factor receptor superfamily, member 6b, decoy; UNQ186/PRO212 |
| Símbolos de los genes | TNFRSF6B |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?