missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DBX2 Polyclonal Antibody

Descripción
A suggested positive control is human kidney tissue lysate. PA5-34391 can be used with blocking peptide PEP-1434.
DBX2 is a member of the developing brain homeobox (DBX) protein family, but while the related protein DBX1 is expressed in various regions of the developing brain, DBX2 shows a more restricted pattern of expression in the brain, and is also expressed in some mesenchymal cells such as limb buds and tooth germs. It is thought that DBX1 and DBX2 promote the development of a subset of interneurons, some of which help mediate left-right coordination of locomotor activity. In Xenopus, DBX2 is involved in primary neurogenesis and early neural plate patterning, and is thought to act as a cross-repressive partner of NKX6-2 in the patterning of the ventral neural tube.
Especificaciones
Especificaciones
| Antígeno | DBX2 |
| Aplicaciones | Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide |
| génica | DBX2 |
| N.º de referencia del gen | Q6ZNG2 |
| Alias de gen | 9430056A22Rik; 9630005K15; AI846217; DBX2; developing brain homeobox 2; developing brain homeobox protein 2; Gm1229; Homeobox protein DBX2 |
| Símbolos de los genes | DBX2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?