missing translation for 'onlineSavingsMsg'
Learn More

Daurisoline, MedChemExpress

Código de producto. 16484787
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16484787 50 mg 50 miligramos
16481338 5 mg 5 miligramos
16478898 10 mg 10 miligramos
16423846 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484787 Supplier Medchem Express Supplier No. HYN0221

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Daurisoline is a hERG inhibitor and also an autophagy blocker.

Daurisoline is a hERG inhibitor and also an autophagy blocker.

Specifications

Nombre del producto químico o material Daurisoline
CAS 70553-76-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C37H42N2O6
Cantidad 50 mg
Sinónimo (R,R)-Daurisoline
Información de solubilidad DMSO : 100 mg/mL (163.74 mM; Need ultrasonic)
SMILES OC1=CC2=C(C=C1OC)CCN(C)[C@@H]2CC3=CC=C(O)C(OC4=CC=C(C[C@H]5N(C)CCC6=C5C=C(OC)C(OC)=C6)C=C4)=C3
Peso molecular (g/mol) 610.74
Formula Weight (peso de la fórmula) 610.74
Porcentaje de pureza 98.45%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.