missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DACT1 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17172064 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
17172064 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17172064

Marca: Invitrogen™ PA5114208

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: LSLVQKKTHP VRTNKPRTSV NADPTKGLLR NGSVCVRAPG GVSQGNSVNL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 85%.

Dapper homolog 1, an ortholog of Xenopus Dpr, antagonizes Wnt signaling by promoting Dvl degradation via a lysosome inhibitor-sensitive and proteasome inhibitor-insensitive mechanism. Dapper homolog 1 acts as an interacting protein for Dvl, and the interaction is formed between the DEP (Dishevelled, Egl-10, and pleckstrin) domain of Dvl and the central and C-terminal regions of Dapper homolog 1. The gene for human Dapper homolog 1 is localized on chromosomal region 14q23.1. The protein impedes the degradation of CTNNB1/beta-catenin, thereby enhancing the transcriptional activation of target genes of the Wnt signaling pathway. The protein is also required for normal notochord formation and is downregulated in hepatocellular carcinomas (HCC). It is expressed in human liver and various other tissues.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno DACT1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica DACT1
N.º de referencia del gen Q9NYF0
Alias de gen 4921528D17Rik; AI115603; dact1; dact1.S; dact1-a; dact1-b; DAPPER; dapper 1; dapper 1-A; dapper antagonist of catenin 1; Dapper homolog 1; dapper homolog 1, antagonist of beta-catenin; dapper homolog 1, antagonist of beta-catenin (xenopus); dapper, antagonist of beta-catenin; dapper, antagonist of beta-catenin, homolog 1; dapper1; dapper1b; dishevelled binding antagonist of beta catenin 1; dishevelled-binding antagonist of beta-catenin 1; dishevelled-binding antagonist of beta-catenin 1 S homeolog; dishevelled-interacting protein; dpr; dpr1; frd1; FRODO; frodo 1; frodo homolog; Frodo1; functional regulator of dsh in ontogenesis; HDPR1; Hepatocellular carcinoma novel gene 3 protein; heptacellular carcinoma novel gene 3; HNG3; MDpr1; MTNG3; RGD1564008; signaling protein; thyex3; thymus expressed gene 3; thymus-expressed novel gene 3 protein; XDpr; XDpr1b; XELAEV_18041122mg
Símbolos de los genes DACT1
Especie del huésped Rabbit
Inmunógeno Immunogen sequence: LSLVQKKTHP VRTNKPRTSV NADPTKGLLR NGSVCVRAPG GVSQGNSVNL.
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 51339
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.