Learn More
DAB2 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16806283 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16806283 20 μL 20 microlitros
16896273 150 μL 150 microlitros
2 options
Código de producto. 16806283
Marca: Proteintech 101092AP20UL
Rabbit Polyclonal Antibody
DAB2 mRNA is expressed in normal ovarian epithelial cells but is down-regulated or absent from ovarian carcinoma cell lines. The 770-amino acid predicted protein has an overall 83% identity with the mouse p96 protein, a putative mitogen-responsive phosphoprotein; homology is strongest in the amino-terminal end of the protein in a region corresponding to the phosphotyrosine interaction domain. The down-regulation of DAB2 may play an important role in ovarian carcinogenesis. This gene was initially named DOC2 (for Differentially expressed in Ovarian Cancer) and is distinct from the DOC2A and DOC2B genes (for double C2-like domains, alpha and beta).
Especificaciones
Antígeno DAB2
Aplicaciones Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence
Clasificación Polyclonal
Concentración 0.37 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica DAB2
N.º de referencia del gen P98082
Alias de gen DAB2, Disabled homolog 2, DOC 2, DOC2
Símbolos de los genes DAB2
Especie del huésped Rabbit
Inmunógeno DAB2 Fusion Protein Ag0162
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1601
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG