missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4-GDI/RhoGDI2 Antibody (97A1015), Alexa Fluor™ 488 - (cleavage specific), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB100-56725AF488
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
D4-GDI/RhoGDI2 Monoclonal specifically detects D4-GDI/RhoGDI2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Especificaciones
| D4-GDI/RhoGDI2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARHGDIB Rho GDP dissociation inhibitor (GDI) beta D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 397 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 97A1015 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
| P52566 | |
| ARHGDIB | |
| A synthetic peptide corresponding to the region of the Fas-induced cleavage site of human D4-GDI/RhoGDI2 was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| In anti-Fas treated Jurkat, an approx. 23 kDa band should be observed. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido