missing translation for 'onlineSavingsMsg'
Learn More

Melford D-(+)-Raffinose Pentahydrate

Código de producto. 30018751
Change view
Click to view available options
Cantidad:
25 g
100 g
1 kg
5 kg
Tamaño de la unidad:
1 Kg
100 g
25 g
5 Kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30018751 25 g 25 g
30018752 5 kg 5 Kg
30018750 1 kg 1 Kg
30018749 100 g 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018751 Supplier Melford Supplier No. R2050025.0

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Sulfated Ash: <0.1%

Heavy Metals: <0.001%

Specifications

CAS 17629-30-0, 512-69-6
Fórmula molecular C18H32O16-5H2O
Cantidad 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.