Learn More
STEMCELL Technologies D-PBS, 10X Concentrate (Without Ca++ and Mg++)
Código de producto. 16645655
Encompass_Preferred
Change view
Click to view available options
Cantidad:
500 mL
Tamaño de la unidad:
paquete de 500
16645655 500 mL paquete de 500
1 options
Código de producto. 16645655
Marca: STEMCELL Technologies 37354
Dulbecco’s phosphate-buffered saline, 10X concentrate, without calcium and magnesium
  • Dulbecco's phosphate-buffered saline (D-PBS) can be used as a temporary diluting, washing, irrigating, or transporting solution for cell or tissue culture
  • It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions
Especificaciones
Cantidad 500 mL