Learn More
Cytokeratin, pan Antibody (AE-1/AE-3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: R&D Systems NBP2-33200AF532
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit, Reptile, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF, Single Cell Western, Dual RNAscope ISH-IHC
Especificaciones
| Cytokeratin, pan | |
| Monoclonal | |
| Alexa Fluor 532 | |
| AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A;NEPPK | |
| Human epidermal keratin | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 3848 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| AE-1/AE-3 | |
| 50 mM Sodium borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Canine | |
| Purified |