missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 7/17 Monoclonal Antibody (C-46)

Descripción
This antibody reacts with cytokeratin peptides 7 and 17 (54 and 46 kDa). Cytokeratins are members of intermediate filaments subfamily of intracellular proteins represented in epithelial tissues. This antibody will not cross-react with mouse, rat or rabbit.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Especificaciones
Especificaciones
| Antígeno | Cytokeratin 7/17 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | C-46 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 15mM sodium azide; pH 7.4 |
| génica | KRT7 |
| N.º de referencia del gen | A1L595, P08729, Q04695, Q29S21 |
| Alias de gen | 39.1; CK-17; CK7; CK-7; cytokeratin 7; cytokeratin-17; Cytokeratin-7; K17; K2C7; K7; keratin 14; keratin 17; keratin 17, type I; keratin 7; keratin 7, type II; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type I cytoskeletal 17; Keratin, type II cytoskeletal 7; keratin-17; Keratin-7; keratin-associated protein 7-1; KRT14; KRT17; KRT7; LOC100626483; LOC100737113; PC; PC2; PCHC1; sarcolectin; SCL; similar to keratin 7; type II mesothelial keratin K7; type-II keratin Kb7 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?